Assay Details
| Gene Symbol: ZNF653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138783.3 | NP_620138.2 | 4 - 5 | 1309 | 63 | |
| GenBank mRNA | AY072704.1 | - | 4 - 5 | 1178 | 63 | |
| BC014187.2 | - | 4 - 5 | 1205 | 4634585 | 63 | |
| BC016816.1 | - | 4 - 5 | 1229 | 4094670 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19002678.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19016548.hg.1 | TC19002678.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016548.hg.1 | ENST00000293771.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19016548.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000593191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000593191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | ENST00000593191.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | TC1900011871.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900204314.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000293771.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000588541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000589051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000590548.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000592756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000593191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000593191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | ENST00000593191.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011871.hg.1 | NM_138783.3 | Refseq |

ZNF653
Genomic Map