Assay Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318230.1 | NP_001305159.1 | 3 - 4 | 731 | 59 | |
| NM_006285.2 | NP_006276.2 | 3 - 4 | 731 | 59 | ||
| GenBank mRNA | AK298907.1 | - | 3 - 4 | 269 | 59 | |
| BC037561.1 | - | 3 - 4 | 326 | 59 | ||
| BC038448.1 | - | 3 - 4 | 705 | 5170888 | 59 | |
| BC067130.1 | - | 3 - 4 | 742 | 6153652 | 59 | |
| D50863.1 | - | 3 - 4 | 667 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000187.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001318230.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NR_039813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002079.hg.1 | TC09000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002079.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002079.hg.1 | ENST00000498522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002079.hg.1 | ENST00000535770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002079.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NR_039813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148130.hg.1 | TC0900007085.hg.1 | Clariom D TC |

TESK1
Genomic Map