Assay Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318230.1 | NP_001305159.1 | 7 - 8 | 1140 | 73 | |
| NM_006285.2 | NP_006276.2 | 8 - 9 | 1223 | 73 | ||
| GenBank mRNA | AF507907.1 | - | 7 - 8 | 636 | 73 | |
| AK298907.1 | - | 8 - 9 | 761 | 73 | ||
| BC037561.1 | - | 8 - 9 | 818 | 73 | ||
| BC038448.1 | - | 8 - 9 | 1197 | 5170888 | 73 | |
| BC067130.1 | - | 8 - 9 | 1234 | 6153652 | 73 | |
| D50863.1 | - | 8 - 9 | 1159 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000187.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09002096.hg.1 | TC09000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002096.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002096.hg.1 | ENST00000498522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002096.hg.1 | ENST00000535770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002096.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007085.hg.1 | NR_039813.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002091.hg.1 | TC09000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002091.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002091.hg.1 | ENST00000498522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002091.hg.1 | ENST00000535770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002091.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002095.hg.1 | TC09000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002095.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002095.hg.1 | ENST00000498522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002095.hg.1 | ENST00000535770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002095.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148149.hg.1 | TC0900007085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148145.hg.1 | TC0900007085.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000463897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000467424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000578933.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007085.hg.1 | NR_039813.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000336395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000480077.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000498522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | ENST00000620767.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | NM_006285.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900148148.hg.1 | TC0900007085.hg.1 | Clariom D TC |

TESK1
Genomic Map