Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
233106_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009166.hg.1 |
ENST00000617151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009166.hg.1 |
NR_037676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012864.hg.1 |
NR_037676.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14012864.hg.1 |
TC14001134.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400167650.hg.1 |
ENST00000617151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167650.hg.1 |
NR_037676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167650.hg.1 |
ENST00000617151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400167650.hg.1 |
NR_037676.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400167650.hg.1 |
TC1400009166.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009166.hg.1 |
ENST00000617151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009166.hg.1 |
NR_037676.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
233106_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
233106_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
233106_at9 |
- |
- |
Human Genome U95 Set |
79914_r_at |
- |
- |
Human Genome U95 Set |
79915_at |
- |
- |
Human Genome U95 Set |
79915_PM_at |
- |
- |
Human Genome U95 Set |
79914_PM_r_at |
- |
- |
Human Genome U133 Set |
233106_PM_at |
- |
- |
Human Genome U133 Set |
233106_at |
- |
- |