Assay Details
| Gene Symbol: NDUFA4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020142.3 | NP_064527.1 | 1 - 2 | 315 | 64 | |
| XM_005269033.2 | XP_005269090.1 | 3 - 4 | 288 | 64 | ||
| XM_011538573.2 | XP_011536875.1 | 2 - 3 | 930 | 64 | ||
| GenBank mRNA | AF164796.1 | - | 1 - 2 | 60 | 64 | |
| AK094275.1 | - | 5 - 6 | 1145 | 64 | ||
| AK129629.1 | - | 1 - 2 | 63 | 64 | ||
| BC011910.2 | - | 1 - 2 | 315 | 4138587 | 64 | |
| CR457176.1 | - | 1 - 2 | 51 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001622.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12021835.hg.1 | ENST00000393825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021835.hg.1 | ENST00000554503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021835.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021835.hg.1 | TC12001622.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021836.hg.1 | ENST00000393825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021836.hg.1 | ENST00000554503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021836.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021836.hg.1 | TC12001622.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000557276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | ENST00000557276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021833.hg.1 | ENST00000393825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021833.hg.1 | ENST00000554503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021833.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021833.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021833.hg.1 | TC12001622.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021834.hg.1 | ENST00000393825.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021834.hg.1 | ENST00000554503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021834.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021834.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021834.hg.1 | TC12001622.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000557276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | ENST00000557276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010961.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184992.hg.1 | TC1200010961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184988.hg.1 | TC1200010961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000554688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000555173.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | ENST00000556732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184987.hg.1 | TC1200010961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184991.hg.1 | TC1200010961.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | ENST00000556234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | ENST00000393825.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | ENST00000554503.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | ENST00000556234.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | NM_020142.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200184994.hg.1 | TC1200010961.hg.1 | Clariom D TC |

NDUFA4L2
Genomic Map