Assay Details
| Gene Symbol: JMJD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282948.1 | NP_001269877.1 | 10 - 11 | 5502 | 132 | |
| NM_001318153.1 | NP_001305082.1 | 10 - 11 | 5506 | 132 | ||
| NM_001318154.1 | NP_001305083.1 | 11 - 12 | 5444 | 132 | ||
| NM_001322252.1 | NP_001309181.1 | 10 - 11 | 5499 | 132 | ||
| NM_001322254.1 | NP_001309183.1 | 9 - 10 | 5396 | 132 | ||
| NM_001322258.1 | NP_001309187.1 | 10 - 11 | 5338 | 132 | ||
| NM_032776.2 | NP_116165.1 | 11 - 12 | 5613 | 132 | ||
| XM_011539503.1 | XP_011537805.1 | 10 - 11 | 5106 | 132 | ||
| XM_017015897.1 | XP_016871386.1 | 10 - 11 | 5101 | 132 | ||
| XM_017015898.1 | XP_016871387.1 | 11 - 12 | 5294 | 132 | ||
| XM_017015899.1 | XP_016871388.1 | 11 - 12 | 5194 | 132 | ||
| XM_017015900.1 | XP_016871389.1 | 11 - 12 | 5598 | 132 | ||
| XM_017015901.1 | XP_016871390.1 | 10 - 11 | 5141 | 132 | ||
| XM_017015902.1 | XP_016871391.1 | 10 - 11 | 5402 | 132 | ||
| XM_017015903.1 | XP_016871392.1 | 9 - 10 | 4950 | 132 | ||
| GenBank mRNA | AB037801.1 | - | 2 - 3 | 1453 | 132 | |
| AK056660.1 | - | 2 - 3 | 1989 | 132 | ||
| AK095584.1 | - | 4 - 5 | 2609 | 132 | ||
| AL831917.1 | - | 4 - 5 | 4141 | 132 | ||
| BC143722.1 | - | 10 - 11 | 5345 | 9052236 | 132 | |
| EF068222.1 | - | 13 - 14 | 5570 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001329.hg.1
Set Membership: |
|

JMJD1C
Genomic Map