Assay Details
| Gene Symbol: JMJD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282948.1 | NP_001269877.1 | 19 - 20 | 7056 | 120 | |
| NM_001318153.1 | NP_001305082.1 | 19 - 20 | 7060 | 120 | ||
| NM_001318154.1 | NP_001305083.1 | 20 - 21 | 6998 | 120 | ||
| NM_001322252.1 | NP_001309181.1 | 19 - 20 | 7053 | 120 | ||
| NM_001322254.1 | NP_001309183.1 | 18 - 19 | 6950 | 120 | ||
| NM_001322258.1 | NP_001309187.1 | 19 - 20 | 6892 | 120 | ||
| NM_032776.2 | NP_116165.1 | 20 - 21 | 7167 | 120 | ||
| NR_134512.1 | 16 - 17 | 6580 | 120 | |||
| XM_011539503.1 | XP_011537805.1 | 19 - 20 | 6660 | 120 | ||
| XM_017015897.1 | XP_016871386.1 | 19 - 20 | 6655 | 120 | ||
| XM_017015898.1 | XP_016871387.1 | 20 - 21 | 6848 | 120 | ||
| XM_017015899.1 | XP_016871388.1 | 20 - 21 | 6748 | 120 | ||
| XM_017015900.1 | XP_016871389.1 | 20 - 21 | 7152 | 120 | ||
| XM_017015901.1 | XP_016871390.1 | 19 - 20 | 6695 | 120 | ||
| XM_017015902.1 | XP_016871391.1 | 19 - 20 | 6956 | 120 | ||
| XM_017015903.1 | XP_016871392.1 | 18 - 19 | 6504 | 120 | ||
| GenBank mRNA | AB037801.1 | - | 11 - 12 | 3004 | 120 | |
| AK024991.1 | - | 9 - 10 | 2467 | 120 | ||
| AL831917.1 | - | 13 - 14 | 5695 | 120 | ||
| BC143722.1 | - | 19 - 20 | 6899 | 9052236 | 120 | |
| BX537954.1 | - | 16 - 17 | 6543 | 120 | ||
| DR762886.1 | - | 3 - 4 | 454 | 120 | ||
| EF068222.1 | - | 22 - 23 | 7124 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001329.hg.1
Set Membership: |
|

JMJD1C
Genomic Map