Assay Details
| Gene Symbol: BACH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170794.1 | NP_001164265.1 | 3 - 4 | 688 | 83 | |
| NM_021813.3 | NP_068585.1 | 5 - 6 | 859 | 83 | ||
| XM_011536039.2 | XP_011534341.1 | 4 - 5 | 770 | 83 | ||
| XM_011536040.2 | XP_011534342.1 | 3 - 4 | 719 | 83 | ||
| XM_017011165.1 | XP_016866654.1 | 4 - 5 | 785 | 83 | ||
| GenBank mRNA | AB208889.1 | - | 3 - 4 | 325 | 83 | |
| AF357835.1 | - | 2 - 3 | 184 | 83 | ||
| AJ271878.1 | - | 5 - 6 | 693 | 83 | ||
| AK309526.1 | - | 5 - 6 | 990 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001946.hg
Additional Information:
This assay is in the 5'UTR region of the AF357835.1,AJ271878.1,NM_001170794.1,NM_021813.3,XM_011536039.2,XM_011536040.2,XM_017011165.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023963.hg.1 | TC06001946.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023963.hg.1 | ENST00000257749.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023963.hg.1 | ENST00000343122.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023963.hg.1 | ENST00000537989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023963.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06023963.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186631.hg.1 | TC0600012554.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |

BACH2
Genomic Map