Assay Details
| Gene Symbol: BACH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021813.3 | NP_068585.1 | 3 - 4 | 601 | 82 | |
| XM_011536039.2 | XP_011534341.1 | 2 - 3 | 512 | 82 | ||
| XM_017011156.1 | XP_016866645.1 | 3 - 4 | 605 | 82 | ||
| XM_017011157.1 | XP_016866646.1 | 3 - 4 | 605 | 82 | ||
| XM_017011161.1 | XP_016866650.1 | 3 - 4 | 605 | 82 | ||
| XM_017011164.1 | XP_016866653.1 | 2 - 3 | 512 | 82 | ||
| XM_017011166.1 | XP_016866655.1 | 3 - 4 | 605 | 82 | ||
| XM_017011167.1 | XP_016866656.1 | 3 - 4 | 605 | 82 | ||
| GenBank mRNA | AA885077.1 | - | 3 - 4 | 431 | 82 | |
| AJ271878.1 | - | 3 - 4 | 435 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ271878.1,NM_021813.3,XM_011536039.2,XM_017011156.1,XM_017011157.1,XM_017011161.1,XM_017011164.1,XM_017011166.1,XM_017011167.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600186640.hg.1 | TC0600012554.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000257749.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000343122.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000406998.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000453877.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000470301.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000472023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000481150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000493201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000494747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | ENST00000537989.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_001170794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012554.hg.1 | NM_021813.3 | Refseq |

BACH2
Genomic Map