Assay Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005858.3 | NP_005849.1 | 10 - 11 | 1371 | 77 | |
| XM_011527624.2 | XP_011525926.1 | 10 - 11 | 1464 | 77 | ||
| XM_017026141.1 | XP_016881630.1 | 10 - 11 | 1516 | 77 | ||
| GenBank mRNA | AK303640.1 | - | 7 - 8 | 998 | 77 | |
| AK308251.1 | - | 9 - 10 | 1265 | 77 | ||
| AK309529.1 | - | 7 - 8 | 899 | 77 | ||
| AL050160.1 | - | 4 - 5 | 299 | 77 | ||
| BC037270.2 | - | 10 - 11 | 1361 | 4837309 | 77 | |
| Y11997.1 | - | 10 - 11 | 1362 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001253.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017619.hg.1 | ENST00000269701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017619.hg.1 | ENST00000537303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017619.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017619.hg.1 | TC19001253.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017620.hg.1 | ENST00000269701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017620.hg.1 | ENST00000537303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017620.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017620.hg.1 | TC19001253.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | TC1900009866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180874.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | TC1900009866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180875.hg.1 | NM_005858.3 | Refseq |

AKAP8
Genomic Map