Assay Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005858.3 | NP_005849.1 | 2 - 3 | 130 | 86 | |
| XM_011527624.2 | XP_011525926.1 | 2 - 3 | 272 | 86 | ||
| XM_017026141.1 | XP_016881630.1 | 2 - 3 | 275 | 86 | ||
| GenBank mRNA | BC037270.2 | - | 2 - 3 | 120 | 4837309 | 86 |
| DR005936.1 | - | 2 - 3 | 133 | 86 | ||
| Y11997.1 | - | 2 - 3 | 121 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001253.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011527624.2,XM_017026141.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017628.hg.1 | ENST00000269701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017628.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017628.hg.1 | TC19001253.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | TC1900009866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180886.hg.1 | NM_005858.3 | Refseq |

AKAP8
Genomic Map