Assay Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005858.3 | NP_005849.1 | 4 - 5 | 439 | 110 | |
| XM_017026141.1 | XP_016881630.1 | 4 - 5 | 584 | 110 | ||
| GenBank mRNA | AK308251.1 | - | 2 - 3 | 176 | 110 | |
| BC037270.2 | - | 4 - 5 | 429 | 4837309 | 110 | |
| DR005936.1 | - | 4 - 5 | 442 | 110 | ||
| Y11997.1 | - | 4 - 5 | 430 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001253.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
mitotic chromosome condensation
signal transduction
protein transport
positive regulation of histone deacetylation
regulation of histone phosphorylation
cell cycle G2/M phase transition
innate immune response
double-stranded DNA binding
protein binding
zinc ion binding
DEAD/H-box RNA helicase binding
protein kinase A regulatory subunit binding
histone deacetylase binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19017626.hg.1 | ENST00000269701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017626.hg.1 | ENST00000537303.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017626.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19017626.hg.1 | TC19001253.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000595416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009866.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | TC1900009866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000598597.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180882.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | NM_005858.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | TC1900009866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | ENST00000269701.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | ENST00000537303.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | ENST00000599883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900180883.hg.1 | NM_005858.3 | Refseq |

AKAP8
Genomic Map