Assay Details
| Gene Symbol: TSHZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308210.1 | NP_001295139.1 | 1 - 2 | 388 | 72 | |
| GenBank mRNA | BQ924585.1 | - | 389 | 72 | ||
| BX447393.2 | - | 1 - 2 | 57 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
multicellular organism development
anterior/posterior pattern specification
middle ear morphogenesis
soft palate development
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
multicellular organism development
anterior/posterior pattern specification
middle ear morphogenesis
soft palate development
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | TC1800007696.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152685.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | TC1800007696.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152687.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560661.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007696.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | TC1800007696.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000322038.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000560918.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | ENST00000584217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152688.hg.1 | NM_005786.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152680.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152680.hg.1 | NM_001308210.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800152680.hg.1 | TC1800007696.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800152680.hg.1 | ENST00000580243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800152680.hg.1 | NM_001308210.1 | Refseq |

TSHZ1
Genomic Map