Assay Details
| Gene Symbol: TULP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020245.4 | NP_064630.2 | 12 - 13 | 3372 | 76 | |
| XM_011535946.1 | XP_011534248.1 | 14 - 15 | 4118 | 76 | ||
| XM_017011063.1 | XP_016866552.1 | 15 - 16 | 4267 | 76 | ||
| XM_017011064.1 | XP_016866553.1 | 16 - 17 | 4432 | 76 | ||
| XM_017011065.1 | XP_016866554.1 | 15 - 16 | 4185 | 76 | ||
| XM_017011066.1 | XP_016866555.1 | 14 - 15 | 4075 | 76 | ||
| XM_017011067.1 | XP_016866556.1 | 14 - 15 | 4168 | 76 | ||
| XM_017011068.1 | XP_016866557.1 | 14 - 15 | 4134 | 76 | ||
| XM_017011069.1 | XP_016866558.1 | 14 - 15 | 4201 | 76 | ||
| XM_017011070.1 | XP_016866559.1 | 13 - 14 | 4049 | 76 | ||
| GenBank mRNA | AF219946.2 | - | 12 - 13 | 3372 | 76 | |
| BE795693.1 | - | 76 | ||||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001132.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000461853.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000613994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013483.hg.1 | TC06001132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013483.hg.1 | ENST00000367097.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06013483.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199844.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199844.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199844.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600199844.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600199844.hg.1 | TC0600014198.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000367097.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000461853.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000613994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | ENST00000616856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_001007466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014198.hg.1 | NM_020245.4 | Refseq |

TULP4
Genomic Map