Assay Details
Gene Symbol: RNF217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286398.2 | NP_001273327.1 | 5 - 6 | 1546 | 144 | |
NR_104440.2 | 6 - 7 | 903 | 144 | |||
NR_136734.1 | 6 - 7 | 1667 | 144 | |||
GenBank mRNA | AK057943.1 | - | 2 - 3 | 272 | 144 | |
AK093417.1 | - | 6 - 7 | 903 | 144 | ||
AK122811.1 | - | 5 - 6 | 1375 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
ligase activity
ubiquitin conjugating enzyme binding
metal ion binding
ubiquitin protein ligase activity
protein polyubiquitination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
ligase activity
ubiquitin conjugating enzyme binding
metal ion binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000359704.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368414.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368415.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368415.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368415.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368415.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368415.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000368415.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000431104.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000431104.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000431104.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000432158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000432158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000432158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000432158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000432158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000432158.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000454842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000454842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000454842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000454842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000454842.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519565.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519799.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519799.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519799.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519799.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000519971.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000521654.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | ENST00000560949.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_001286398.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NM_152553.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009350.hg.1 | NR_104440.2 | Refseq |
