Assay Details
| Gene Symbol: AMN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030943.3 | NP_112205.2 | 8 - 9 | 873 | 76 | |
| XM_011537202.2 | XP_011535504.1 | 8 - 9 | 1118 | 76 | ||
| XM_011537203.2 | XP_011535505.1 | 8 - 9 | 1374 | 76 | ||
| GenBank mRNA | AF328788.1 | - | 8 - 9 | 862 | 76 | |
| AK055802.1 | - | 7 - 8 | 1586 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000790.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14008536.hg.1 | ENST00000299155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008536.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008536.hg.1 | TC14000790.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008535.hg.1 | ENST00000299155.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008535.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14008535.hg.1 | TC14000790.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | ENST00000559789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559507.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | ENST00000559789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010690.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180782.hg.1 | TC1400010690.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | ENST00000299155.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | ENST00000541086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | NM_030943.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400180783.hg.1 | TC1400010690.hg.1 | Clariom D TC |

AMN
Genomic Map