Assay Details
| Gene Symbol: NABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024068.3 | NP_076973.1 | 5 - 6 | 495 | 78 | |
| XM_005269146.2 | XP_005269203.1 | 5 - 6 | 506 | 78 | ||
| XM_005269147.2 | XP_005269204.1 | 5 - 6 | 515 | 78 | ||
| XM_005269148.4 | XP_005269205.1 | 5 - 6 | 490 | 78 | ||
| XM_005269149.4 | XP_005269206.1 | 5 - 6 | 555 | 78 | ||
| GenBank mRNA | AY203942.1 | - | 5 - 6 | 454 | 78 | |
| BC001079.2 | - | 4 - 5 | 505 | 2822460 | 78 | |
| BC006171.1 | - | 5 - 6 | 469 | 4026297 | 78 | |
| BG697807.1 | - | 5 - 6 | 497 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000507.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA repair
cellular response to DNA damage stimulus
mitotic cell cycle checkpoint
response to ionizing radiation
snRNA transcription from RNA polymerase II promoter
regulation of telomerase activity
establishment of protein localization to telomere
positive regulation of telomere capping
single-stranded DNA binding
protein binding
enzyme binding
C-rich strand telomeric DNA binding
G-rich strand telomeric DNA binding
double-strand break repair via homologous recombination
DNA repair
cellular response to DNA damage stimulus
mitotic cell cycle checkpoint
response to ionizing radiation
snRNA transcription from RNA polymerase II promoter
regulation of telomerase activity
establishment of protein localization to telomere
positive regulation of telomere capping
single-stranded DNA binding
protein binding
enzyme binding
C-rich strand telomeric DNA binding
G-rich strand telomeric DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006648.hg.1 | ENST00000267023.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006648.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006648.hg.1 | ENST00000380198.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006648.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006648.hg.1 | TC12000507.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200156858.hg.1 | TC1200007832.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000267023.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000341463.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000380198.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000399713.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000447747.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | ENST00000479016.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007832.hg.1 | NM_024068.3 | Refseq |

NABP2
Genomic Map