Assay Details
| Gene Symbol: SLC30A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004434.2 | NP_001004434.1 | 4 - 5 | 817 | 119 | |
| NM_032513.4 | NP_115902.1 | 3 - 4 | 670 | 119 | ||
| GenBank mRNA | AF370409.1 | - | 4 - 5 | 664 | 119 | |
| AK094027.1 | - | 4 - 5 | 633 | 119 | ||
| BC006251.2 | - | 3 - 4 | 645 | 3948165 | 119 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002385.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01036767.hg.1 | TC01002385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036767.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036767.hg.1 | ENST00000374278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036767.hg.1 | ENST00000486732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036767.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036767.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036768.hg.1 | TC01002385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036768.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036768.hg.1 | ENST00000374278.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036768.hg.1 | ENST00000486732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036768.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036768.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | ENST00000498060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013373.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199341.hg.1 | TC0100013373.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | ENST00000374276.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | ENST00000374278.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | NM_001004434.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | NM_032513.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199342.hg.1 | TC0100013373.hg.1 | Clariom D TC |

SLC30A2
Genomic Map