Assay Details
| Gene Symbol: PDGFRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006207.2 | NP_006198.1 | 2 - 3 | 501 | 64 | |
| GenBank mRNA | AK289450.1 | - | 2 - 3 | 501 | 64 | |
| BC010927.1 | - | 1 - 2 | 72 | 4278882 | 64 | |
| CR541952.1 | - | 1 - 2 | 56 | 64 | ||
| D37965.1 | - | 1 - 2 | 117 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000128.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08001225.hg.1 | TC08000128.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001225.hg.1 | ENST00000251630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001225.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001225.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001228.hg.1 | TC08000128.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001228.hg.1 | ENST00000251630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001228.hg.1 | ENST00000398074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001228.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001228.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001226.hg.1 | TC08000128.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001226.hg.1 | ENST00000251630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001226.hg.1 | ENST00000398074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001226.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001226.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001227.hg.1 | TC08000128.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001227.hg.1 | ENST00000251630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001227.hg.1 | ENST00000398074.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001227.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001227.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000523248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000523248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001223.hg.1 | TC08000128.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001223.hg.1 | ENST00000251630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001223.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08001223.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146525.hg.1 | TC0800006873.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000523248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000523248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006873.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146523.hg.1 | TC0800006873.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146529.hg.1 | TC0800006873.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | ENST00000251630.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | ENST00000541323.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | NM_006207.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800146527.hg.1 | TC0800006873.hg.1 | Clariom D TC |

PDGFRL
Genomic Map