Assay Details
Gene Symbol: KRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007043.6 | NP_008974.5 | 4 - 5 | 566 | 96 | |
GenBank mRNA | AK313687.1 | - | 4 - 5 | 529 | 96 | |
BC005225.1 | - | 4 - 5 | 561 | 96 | ||
BC014034.2 | - | 4 - 5 | 542 | 96 | ||
BC016778.1 | - | 4 - 5 | 540 | 4106298 | 96 | |
BC026107.2 | - | 4 - 5 | 549 | 4827704 | 96 | |
BC033887.2 | - | 4 - 5 | 540 | 5268517 | 96 | |
U55766.1 | - | 4 - 5 | 537 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001747.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12023118.hg.1 | ENST00000229214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12023118.hg.1 | ENST00000438169.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12023118.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12023118.hg.1 | TC12001747.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12023120.hg.1 | ENST00000229214.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12023120.hg.1 | ENST00000438169.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12023120.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12023120.hg.1 | TC12001747.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000551070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1200011301.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187613.hg.1 | TC1200011301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000550898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187614.hg.1 | TC1200011301.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000229214.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000438169.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000550023.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | ENST00000612627.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | NM_007043.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200187611.hg.1 | TC1200011301.hg.1 | Clariom D TC |
