Assay Details
| Gene Symbol: ZFAND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021943.2 | NP_068762.1 | 4 - 5 | 777 | 86 | |
| XM_017011171.1 | XP_016866660.1 | 3 - 4 | 750 | 86 | ||
| GenBank mRNA | AK023284.1 | - | 4 - 5 | 451 | 86 | |
| BC031481.2 | - | 4 - 5 | 393 | 4448968 | 86 | |
| BP305887.1 | - | 4 - 5 | 506 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000549.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06006059.hg.1 | TC06000549.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006059.hg.1 | ENST00000287218.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006059.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000469208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000469208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006061.hg.1 | TC06000549.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006061.hg.1 | ENST00000287218.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006061.hg.1 | ENST00000373391.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06006061.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154814.hg.1 | TC0600007887.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154811.hg.1 | TC0600007887.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000287218.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000373391.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000440482.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000463847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000469208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000469208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | ENST00000474522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007887.hg.1 | NM_021943.2 | Refseq |

ZFAND3
Genomic Map