Assay Details
| Gene Symbol: EHMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318833.1 | NP_001305762.1 | 10 - 11 | 1471 | 108 | |
| NM_006709.4 | NP_006700.3 | 10 - 11 | 1242 | 108 | ||
| XM_005248824.3 | XP_005248881.1 | 9 - 10 | 1383 | 108 | ||
| XM_006714974.1 | XP_006715037.1 | 9 - 10 | 1387 | 108 | ||
| XM_006714976.1 | XP_006715039.1 | 10 - 11 | 1225 | 108 | ||
| GenBank mRNA | AB209433.2 | - | 9 - 10 | 1304 | 108 | |
| AJ315532.1 | - | 10 - 11 | 1212 | 108 | ||
| BC002686.1 | - | 7 - 8 | 712 | 108 | ||
| BC009351.1 | - | 10 - 11 | 1209 | 108 | ||
| BC018718.1 | - | 10 - 11 | 1200 | 4123580 | 108 | |
| BC020970.1 | - | 9 - 10 | 1177 | 108 | ||
| X69838.1 | - | 6 - 7 | 632 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

EHMT2
Genomic Map