Assay Details
| Gene Symbol: DHRS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005266528.4 | XP_005266585.1 | 7 - 8 | 716 | 83 | |
| XM_017020749.1 | XP_016876238.1 | 8 - 9 | 785 | 83 | ||
| XM_017020752.1 | XP_016876241.1 | 6 - 7 | 595 | 83 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000218981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000280056.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000444610.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000461948.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000469957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000472372.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000489680.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000490949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | ENST00000497151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001031719.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_001270424.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008997.hg.1 | NM_024705.2 | Refseq |

DHRS12
Genomic Map