Assay Details
| Gene Symbol: MAP2K5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206804.1 | NP_001193733.1 | 19 - 20 | 1205 | 62 | |
| NM_002757.3 | NP_002748.1 | 18 - 19 | 1759 | 62 | ||
| NM_145160.2 | NP_660143.1 | 19 - 20 | 1789 | 62 | ||
| GenBank mRNA | AK024766.1 | - | 8 - 9 | 383 | 62 | |
| AK057521.1 | - | 13 - 14 | 917 | 62 | ||
| AK293459.1 | - | 19 - 20 | 1205 | 62 | ||
| AK315435.1 | - | 19 - 20 | 1336 | 62 | ||
| BC008838.2 | - | 19 - 20 | 1446 | 4100679 | 62 | |
| BT006780.1 | - | 19 - 20 | 1136 | 62 | ||
| CR542229.1 | - | 19 - 20 | 1136 | 62 | ||
| U71087.1 | - | 19 - 20 | 1197 | 62 | ||
| U71088.2 | - | 18 - 19 | 1111 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000625.hg.1
Set Membership: |
|

MAP2K5
Genomic Map