Assay Details
| Gene Symbol: URB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014825.2 | NP_055640.2 | 37 - 38 | 6125 | 77 | |
| GenBank mRNA | AB011111.2 | - | 32 - 33 | 5359 | 77 | |
| AF231919.1 | - | 37 - 38 | 5963 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000397.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000492603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | ENST00000492603.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21004369.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21004369.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21004369.hg.1 | TC21000397.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000480196.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000492603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | ENST00000492603.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007952.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100153495.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100153495.hg.1 | NM_014825.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100153495.hg.1 | TC2100007952.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100153495.hg.1 | ENST00000382751.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100153495.hg.1 | NM_014825.2 | Refseq |

URB1
Genomic Map