Assay Details
| Gene Symbol: TEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003215.2 | NP_003206.2 | 14 - 15 | 1560 | 71 | |
| XM_011513737.2 | XP_011512039.1 | 14 - 15 | 1541 | 71 | ||
| XM_017008569.1 | XP_016864058.1 | 13 - 14 | 2766 | 71 | ||
| GenBank mRNA | BC101711.1 | - | 14 - 15 | 1568 | 8069217 | 71 |
| BC101713.1 | - | 14 - 15 | 1568 | 8069219 | 71 | |
| BC143487.1 | - | 14 - 15 | 1568 | 9051995 | 71 | |
| D29767.1 | - | 14 - 15 | 1586 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001176.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
regulation of platelet activation
peptidyl-tyrosine phosphorylation
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
Fc-epsilon receptor signaling pathway
regulation of cell proliferation
tissue regeneration
innate immune response
positive regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
phospholipid binding
metal ion binding
adaptive immune response
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
regulation of platelet activation
peptidyl-tyrosine phosphorylation
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
Fc-epsilon receptor signaling pathway
regulation of cell proliferation
tissue regeneration
innate immune response
positive regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
phospholipid binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04016162.hg.1 | TC04001176.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016162.hg.1 | ENST00000381501.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016162.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04016162.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010618.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000381501.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000505452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000511150.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000511471.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | ENST00000515146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | NM_003215.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400172456.hg.1 | TC0400010618.hg.1 | Clariom D TC |

TEC
Genomic Map