Assay Details
| Gene Symbol: LRPPRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133259.3 | NP_573566.2 | 19 - 20 | 2024 | 85 | |
| XM_006711915.2 | XP_006711978.1 | 19 - 20 | 2033 | 85 | ||
| XM_006711916.3 | XP_006711979.1 | 19 - 20 | 2008 | 85 | ||
| XM_011532473.1 | XP_011530775.1 | 19 - 20 | 2008 | 85 | ||
| XM_017003117.1 | XP_016858606.1 | 19 - 20 | 2006 | 85 | ||
| GenBank mRNA | AK125781.1 | - | 19 - 20 | 1995 | 85 | |
| AK290016.1 | - | 19 - 20 | 1995 | 85 | ||
| AK299869.1 | - | 18 - 19 | 1796 | 85 | ||
| AY289212.1 | - | 19 - 20 | 2024 | 85 | ||
| BC050311.1 | - | 19 - 20 | 1975 | 4827456 | 85 | |
| BC130285.1 | - | 19 - 20 | 2020 | 40146316 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001795.hg.1
Set Membership: |
|

LRPPRC
Genomic Map