Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016481.4 | NP_057565.3 | 1 - 2 | 149 | 104 | |
| XM_005252047.3 | XP_005252104.1 | 1 - 2 | 106 | 104 | ||
| XM_005252048.4 | XP_005252105.1 | 1 - 2 | 106 | 104 | ||
| XM_011518776.2 | XP_011517078.1 | 1 - 2 | 106 | 104 | ||
| GenBank mRNA | AK000213.1 | - | 1 - 2 | 149 | 104 | |
| AK302701.1 | - | 1 - 2 | 149 | 104 | ||
| AK309270.1 | - | 1 - 2 | 108 | 104 | ||
| AY189280.1 | - | 1 - 2 | 96 | 104 | ||
| BC002863.2 | - | 1 - 2 | 129 | 3944148 | 104 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001400.hg
Additional Information:
This assay is in the 5'UTR region of the AY189280.1,XM_005252048.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09017417.hg.1 | TC09001400.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017417.hg.1 | ENST00000325350.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017417.hg.1 | ENST00000375119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017417.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017418.hg.1 | TC09001400.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017418.hg.1 | ENST00000325350.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017418.hg.1 | ENST00000375119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017418.hg.1 | ENST00000478126.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017418.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017423.hg.1 | TC09001400.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017423.hg.1 | ENST00000375119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017423.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017422.hg.1 | TC09001400.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017422.hg.1 | ENST00000325350.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017422.hg.1 | ENST00000375119.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017422.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174674.hg.1 | TC0900010958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174673.hg.1 | TC0900010958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174680.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174680.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174680.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174680.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174680.hg.1 | TC0900010958.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |

TRMO
Genomic Map