Assay Details
| Gene Symbol: TRMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005252048.4 | XP_005252105.1 | 3 - 4 | 442 | 59 | |
| XM_005252050.4 | XP_005252107.1 | 2 - 3 | 268 | 59 | ||
| XM_011518777.1 | XP_011517079.1 | 2 - 3 | 268 | 59 | ||
| GenBank mRNA | AK302701.1 | - | 3 - 4 | 485 | 59 | |
| AY189280.1 | - | 4 - 5 | 481 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY189280.1, XM_005252048.4, XM_005252050.4, XM_011518777.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375117.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375118.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000375119.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000455506.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000471580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000478126.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | ENST00000611338.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010958.hg.1 | NM_016481.4 | Refseq |

TRMO
Genomic Map