Assay Details
| Gene Symbol: SENP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308045.1 | NP_001294974.1 | 7 - 8 | 2224 | 64 | |
| NM_152699.4 | NP_689912.2 | 8 - 9 | 2362 | 64 | ||
| XM_011512544.2 | XP_011510846.1 | 9 - 10 | 2407 | 64 | ||
| XM_017005870.1 | XP_016861359.1 | 8 - 9 | 2224 | 64 | ||
| GenBank mRNA | AF335474.1 | - | 7 - 8 | 1219 | 64 | |
| AK124389.1 | - | 7 - 8 | 1335 | 64 | ||
| AK302305.1 | - | 7 - 8 | 2224 | 64 | ||
| AK313764.1 | - | 8 - 9 | 2335 | 64 | ||
| BC000958.2 | - | 7 - 8 | 1459 | 64 | ||
| BC008589.1 | - | 7 - 8 | 1459 | 64 | ||
| BC030705.2 | - | 8 - 9 | 2311 | 4824344 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

SENP5
Genomic Map