Assay Details
| Gene Symbol: LRRC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256409.1 | NP_001243338.1 | 5 - 6 | 995 | 79 | |
| NM_052940.4 | NP_443172.1 | 4 - 5 | 961 | 79 | ||
| XM_006710327.3 | XP_006710390.1 | 5 - 6 | 817 | 79 | ||
| XM_006710328.3 | XP_006710391.1 | 5 - 6 | 1330 | 79 | ||
| GenBank mRNA | AK074521.1 | - | 4 - 5 | 934 | 79 | |
| AK075201.1 | - | 4 - 5 | 923 | 79 | ||
| AK090881.1 | - | 5 - 6 | 1244 | 79 | ||
| AK095767.1 | - | 5 - 6 | 914 | 79 | ||
| BC013101.1 | - | 4 - 5 | 800 | 3918785 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000659.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010713.hg.1 | TC01000659.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010713.hg.1 | ENST00000319223.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010713.hg.1 | ENST00000371370.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010713.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010713.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010713.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371368.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000444987.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008369.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | ENST00000319223.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | ENST00000371370.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | ENST00000477905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | NM_001256409.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | NM_052940.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100159612.hg.1 | TC0100008369.hg.1 | Clariom D TC |

LRRC42
Genomic Map