Assay Details
| Gene Symbol: EXOC3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178516.3 | NP_848611.2 | 4 - 5 | 673 | 81 | |
| GenBank mRNA | AK092858.1 | - | 4 - 5 | 565 | 81 | |
| AK292004.1 | - | 4 - 5 | 582 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600181733.hg.1 | TC1600010618.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |

EXOC3L1
Genomic Map