Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282683.1 | NP_001269612.1 | 7 - 8 | 640 | 80 | |
| NM_024735.4 | NP_079011.3 | 6 - 7 | 892 | 80 | ||
| GenBank mRNA | AF318348.1 | - | 5 - 6 | 1092 | 80 | |
| AF428140.1 | - | 6 - 7 | 856 | 80 | ||
| AL117444.1 | - | 4 - 5 | 400 | 80 | ||
| BC012748.1 | - | 6 - 7 | 866 | 3958783 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001330.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
cellular response to DNA damage stimulus
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
mitotic G1 DNA damage checkpoint
positive regulation of dendrite morphogenesis
positive regulation of neuron migration
ubiquitin-protein transferase activity
cyclin binding
protein polyubiquitination
cellular response to DNA damage stimulus
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
mitotic G1 DNA damage checkpoint
positive regulation of dendrite morphogenesis
positive regulation of neuron migration
ubiquitin-protein transferase activity
cyclin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16018358.hg.1 | ENST00000311635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018358.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018358.hg.1 | TC16001330.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000563956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000561664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000563956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000565593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000567622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000569412.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011578.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | ENST00000311635.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | ENST00000563113.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | ENST00000618298.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | NM_001282683.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | NM_024735.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600194755.hg.1 | TC1600011578.hg.1 | Clariom D TC |

FBXO31
Genomic Map