Assay Details
| Gene Symbol: B4GALT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199873.1 | NP_001186802.1 | 2 - 3 | 688 | 151 | |
| XM_017002714.1 | XP_016858203.1 | 2 - 3 | 244 | 151 | ||
| GenBank mRNA | AF038661.1 | - | 2 - 3 | 194 | 151 | |
| AK291455.1 | - | 2 - 3 | 221 | 151 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF038661.1, AK291455.1, NM_001199873.1, XM_017002714.1 transcripts.
Set Membership: |
|

B4GALT3
Genomic Map