Assay Details
| Gene Symbol: BAK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001188.3 | NP_001179.1 | 4 - 5 | 647 | 69 | |
| XM_011514779.2 | XP_011513081.1 | 5 - 6 | 758 | 69 | ||
| XM_011514780.1 | XP_011513082.1 | 4 - 5 | 568 | 69 | ||
| GenBank mRNA | AF520590.1 | - | 3 - 4 | 284 | 69 | |
| AK091807.1 | - | 4 - 5 | 512 | 69 | ||
| AK292093.1 | - | 5 - 6 | 685 | 69 | ||
| AK293547.1 | - | 4 - 5 | 577 | 69 | ||
| BC004431.1 | - | 1 | 558 | 2819507 | 69 | |
| CR457419.1 | - | 3 - 4 | 347 | 69 | ||
| KU177934.1 | - | 3 - 4 | 347 | 69 | ||
| KU177935.1 | - | 3 - 4 | 347 | 69 | ||
| U16811.1 | - | 4 - 5 | 547 | 69 | ||
| U23765.1 | - | 4 - 5 | 411 | 69 | ||
| X84213.1 | - | 4 - 5 | 539 | 69 | ||
More Information
Set Membership: |
|

BAK1
Genomic Map