Assay Details
| Gene Symbol: NSDHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129765.1 | NP_001123237.1 | 2 - 3 | 214 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001129765.1 transcripts.
Set Membership: |
|

NSDHL
Genomic Map