Assay Details
| Gene Symbol: NSDHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129765.1 | NP_001123237.1 | 7 - 8 | 948 | 73 | |
| NM_015922.2 | NP_057006.1 | 6 - 7 | 881 | 73 | ||
| XM_011531178.2 | XP_011529480.1 | 8 - 9 | 1008 | 73 | ||
| XM_017029564.1 | XP_016885053.1 | 6 - 7 | 813 | 73 | ||
| GenBank mRNA | AK026549.1 | - | 6 - 7 | 771 | 73 | |
| BC000245.1 | - | 6 - 7 | 802 | 3355755 | 73 | |
| BC007816.2 | - | 6 - 7 | 852 | 4299819 | 73 | |
| U47105.2 | - | 6 - 7 | 881 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000743.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
hair follicle development
cholesterol biosynthetic process
smoothened signaling pathway
oxidation-reduction process
labyrinthine layer blood vessel development
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
3-beta-hydroxy-delta5-steroid dehydrogenase activity
sterol-4-alpha-carboxylate 3-dehydrogenase (decarboxylating) activity
hair follicle development
cholesterol biosynthetic process
smoothened signaling pathway
oxidation-reduction process
labyrinthine layer blood vessel development
C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
3-beta-hydroxy-delta5-steroid dehydrogenase activity
sterol-4-alpha-carboxylate 3-dehydrogenase (decarboxylating) activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009615.hg.1 | TC0X000743.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009615.hg.1 | ENST00000370274.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009615.hg.1 | ENST00000440023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009615.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X009615.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00159604.hg.1 | TC0X00008759.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000370274.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000432467.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | ENST00000440023.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_001129765.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008759.hg.1 | NM_015922.2 | Refseq |

NSDHL
Genomic Map