Assay Details
| Gene Symbol: CDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016279.3 | NP_057363.3 | 5 - 6 | 980 | 70 | |
| GenBank mRNA | AB035302.1 | - | 5 - 6 | 965 | 70 | |
| AK307779.1 | - | 5 - 6 | 981 | 70 | ||
| AK308697.1 | - | 5 - 6 | 984 | 70 | ||
| BC107594.1 | - | 5 - 6 | 942 | 40011942 | 70 | |
| BC113745.1 | - | 5 - 6 | 942 | 40011941 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001216.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05017167.hg.1 | TC05001216.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017167.hg.1 | ENST00000231021.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05017167.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500172024.hg.1 | TC0500010342.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000231021.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000505045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000511822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | ENST00000513289.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010342.hg.1 | NM_016279.3 | Refseq |

CDH9
Genomic Map