Assay Details
| Gene Symbol: PARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136594.1 | 20 - 21 | 2251 | 59 | ||
| NR_136596.1 | 19 - 20 | 2273 | 59 | |||
| NR_136599.1 | 19 - 20 | 1963 | 59 | |||
| NR_136603.1 | 19 - 20 | 1954 | 59 | |||
| NR_136605.1 | 18 - 19 | 1960 | 59 | |||
| NR_136606.1 | 19 - 20 | 2182 | 59 | |||
| NR_136607.1 | 19 - 20 | 2130 | 59 | |||
| NR_136608.1 | 19 - 20 | 2020 | 59 | |||
| NR_136609.1 | 18 - 19 | 2020 | 59 | |||
| NR_136610.1 | 19 - 20 | 2133 | 59 | |||
| NR_136611.1 | 19 - 20 | 2070 | 59 | |||
| GenBank mRNA | AB499727.1 | - | 16 - 17 | 1488 | 59 | |
| AK022547.1 | - | 18 - 19 | 1714 | 59 | ||
| AK024703.1 | - | 7 - 8 | 558 | 59 | ||
| AK126416.1 | - | 15 - 16 | 2143 | 59 | ||
| AL390093.1 | - | 6 - 7 | 408 | 59 | ||
| BC015176.1 | - | 3 - 4 | 155 | 59 | ||
| BC026955.1 | - | 15 - 16 | 1285 | 59 | ||
More Information
Set Membership: |
|

PARP6
Genomic Map