Assay Details
| Gene Symbol: SLC37A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320537.1 | NP_001307466.1 | 3 - 4 | 1184 | 125 | |
| NM_018964.3 | NP_061837.3 | 4 - 5 | 553 | 125 | ||
| XM_017028377.1 | XP_016883866.1 | 4 - 5 | 478 | 125 | ||
| XM_017028378.1 | XP_016883867.1 | 4 - 5 | 937 | 125 | ||
| XM_017028379.1 | XP_016883868.1 | 5 - 6 | 584 | 125 | ||
| XM_017028380.1 | XP_016883869.1 | 3 - 4 | 384 | 125 | ||
| XM_017028381.1 | XP_016883870.1 | 4 - 5 | 478 | 125 | ||
| XM_017028383.1 | XP_016883872.1 | 4 - 5 | 409 | 125 | ||
| GenBank mRNA | AF311320.1 | - | 3 - 4 | 1123 | 125 | |
| AJ269529.2 | - | 4 - 5 | 553 | 125 | ||
| DA755019.1 | - | 4 - 5 | 478 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000202.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002098.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002098.hg.1 | ENST00000398341.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002098.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21002098.hg.1 | TC21000202.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000398343.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000471277.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000484887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000487951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | ENST00000496416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008527.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | TC2100008527.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | NM_018964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | ENST00000352133.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | ENST00000398341.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100158337.hg.1 | NM_018964.3 | Refseq |

SLC37A1
Genomic Map