Assay Details
| Gene Symbol: GSDMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165959.1 | NP_001159431.1 | 6 - 7 | 821 | 110 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17002851.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | TC17002851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000309481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000360317.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000394175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000394179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000418519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000468820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | ENST00000520542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | NM_001042471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | NM_001165958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | NM_001165959.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019203.hg.1 | NM_018530.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | TC17002851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000309481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000360317.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000394175.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000394179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000418519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000468820.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | ENST00000520542.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | NM_001042471.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | NM_001165958.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | NM_001165959.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17019204.hg.1 | NM_018530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000309481.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000360317.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000394175.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000394179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000418519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000468820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000477054.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000479136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000520542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000522564.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000523371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000524039.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_001042471.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_001165958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_001165959.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_018530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000309481.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000360317.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000394175.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000394179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000418519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000468820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000477054.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000479136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000520542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000522564.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000523371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | ENST00000524039.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_001042471.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_001165958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_001165959.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | NM_018530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183453.hg.1 | TC1700010590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000309481.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000360317.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000394175.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000394179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000418519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000468820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000477054.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000479136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000519429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000520542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000522564.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000523371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000524039.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_001042471.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_001165958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_001165959.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_018530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000309481.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000360317.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000394175.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000394179.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000418519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000468820.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000477054.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000479136.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000519429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000520542.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000522564.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000523371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | ENST00000524039.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_001042471.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_001165958.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_001165959.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | NM_018530.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700183454.hg.1 | TC1700010590.hg.1 | Clariom D TC |

GSDMB
Genomic Map