Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PHD finger protein 20-like 1 | PHF20L1 |
CGI-72, TDRD20B, URLC1 |
-193.9kb | Anti-sense | Chr. 8 - 132775358 - 132848807 on NCBI Build GRCh38 |
| Downstream | microRNA 7848 | MIR7848 | hsa-mir-7848 | 1.5kb | Sense | Chr. 8 - 133046481 - 133046581 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map