Assay Details
| Gene Symbol: RALGAPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020343.3 | NP_065076.2 | 16 - 17 | 2346 | 78 | |
| XM_005260768.2 | XP_005260825.1 | 16 - 17 | 2361 | 78 | ||
| GenBank mRNA | AK002211.1 | - | 8 - 9 | 1216 | 78 | |
| DQ310704.1 | - | 16 - 17 | 2203 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000685.hg.1
Set Membership: |
|

RALGAPA2
Genomic Map