Assay Details
| Gene Symbol: TK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172644.1 | NP_001166115.1 | 2 - 3 | 509 | 77 | |
| GenBank mRNA | AK295808.1 | - | 2 - 3 | 219 | 77 | |
| AK316226.1 | - | 2 - 3 | 219 | 77 | ||
More Information
Set Membership: |
|

TK2
Genomic Map