Assay Details
| Gene Symbol: PPP4R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058237.1 | NP_478144.1 | 18 - 19 | 2203 | 155 | |
| XM_005267930.2 | XP_005267987.1 | 18 - 19 | 2036 | 155 | ||
| XM_011537038.2 | XP_011535340.1 | 18 - 19 | 2199 | 155 | ||
| XM_011537039.2 | XP_011535341.1 | 17 - 18 | 1944 | 155 | ||
| XM_011537040.2 | XP_011535342.1 | 16 - 17 | 1851 | 155 | ||
| XM_017021528.1 | XP_016877017.1 | 16 - 17 | 1888 | 155 | ||
| XM_017021529.1 | XP_016877018.1 | 20 - 21 | 2558 | 155 | ||
| XM_017021530.1 | XP_016877019.1 | 20 - 21 | 2532 | 155 | ||
| GenBank mRNA | AB046842.1 | - | 18 - 19 | 2122 | 155 | |
| AK303722.1 | - | 6 - 7 | 685 | 155 | ||
| BC068491.1 | - | 18 - 19 | 2162 | 5297335 | 155 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000585.hg.1
Set Membership: |
|

PPP4R4
Genomic Map