Assay Details
| Gene Symbol: FYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153047.3 | NP_694592.1 | 6 - 7 | 864 | 111 | |
| XM_005266890.3 | XP_005266947.1 | 9 - 10 | 1041 | 111 | ||
| GenBank mRNA | AK056699.1 | - | 7 - 8 | 1046 | 111 | |
| S74774.1 | - | 6 - 7 | 853 | 111 | ||
More Information
Set Membership: |
|

FYN
Genomic Map