Assay Details
| Gene Symbol: SYNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182961.3 | NP_892006.3 | 137 - 138 | 25582 | 100 | |
| XM_006715413.2 | XP_006715476.1 | 137 - 138 | 25499 | 100 | ||
| XM_006715415.2 | XP_006715478.1 | 137 - 138 | 25499 | 100 | ||
| XM_006715416.2 | XP_006715479.1 | 137 - 138 | 25484 | 100 | ||
| XM_006715425.2 | XP_006715488.1 | 137 - 138 | 25499 | 100 | ||
| XM_017010613.1 | XP_016866102.1 | 137 - 138 | 25496 | 100 | ||
| XM_017010615.1 | XP_016866104.1 | 136 - 137 | 25343 | 100 | ||
| XM_017010616.1 | XP_016866105.1 | 137 - 138 | 25499 | 100 | ||
| XM_017010617.1 | XP_016866106.1 | 137 - 138 | 25496 | 100 | ||
| XM_017010618.1 | XP_016866107.1 | 137 - 138 | 25484 | 100 | ||
| GenBank mRNA | AB018339.2 | - | 25 - 26 | 100 | ||
| AF444779.1 | - | 12 - 13 | 100 | |||
| AF495910.1 | - | 137 - 138 | 100 | |||
| AK122691.1 | - | 17 - 18 | 2862 | 100 | ||
| AK304825.1 | - | 11 - 12 | 1784 | 100 | ||
| AK316101.1 | - | 10 - 11 | 1799 | 100 | ||
| AY061755.1 | - | 52 - 53 | 100 | |||
| AY184206.1 | - | 50 - 51 | 100 | |||
| BC090927.1 | - | 10 - 11 | 1844 | 100 | ||
| BX537837.1 | - | 19 - 20 | 3160 | 100 | ||
More Information
Set Membership: |
|

SYNE1
Genomic Map