Assay Details
| Gene Symbol: RFX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173560.3 | NP_775831.2 | 17 - 18 | 2456 | 78 | |
| XM_011535589.1 | XP_011533891.1 | 16 - 17 | 2343 | 78 | ||
| XM_017010477.1 | XP_016865966.1 | 16 - 17 | 2263 | 78 | ||
| GenBank mRNA | BC039248.1 | - | 17 - 18 | 2456 | 5272028 | 78 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000926.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
type B pancreatic cell differentiation
pancreatic A cell differentiation
pancreatic D cell differentiation
transcription from RNA polymerase II promoter
endocrine pancreas development
positive regulation of insulin secretion involved in cellular response to glucose stimulus
glucose homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin secretion
pancreatic epsilon cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
transcription regulatory region DNA binding
type B pancreatic cell differentiation
pancreatic A cell differentiation
pancreatic D cell differentiation
transcription from RNA polymerase II promoter
endocrine pancreas development
positive regulation of insulin secretion involved in cellular response to glucose stimulus
glucose homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin secretion
pancreatic epsilon cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010864.hg.1 | TC06000926.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010864.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010864.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163513.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163513.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163513.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163513.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163513.hg.1 | TC0600009243.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000332958.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000471966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | ENST00000487683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009243.hg.1 | NM_173560.3 | Refseq |

RFX6
Genomic Map