Assay Details
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018066.3 | NP_060536.3 | 1 - 2 | 699 | 80 | |
GenBank mRNA | AK001211.1 | - | 1 - 2 | 618 | 80 | |
AK022535.1 | - | 1 - 2 | 604 | 80 | ||
AY358864.1 | - | 1 - 2 | 608 | 80 | ||
AY364261.1 | - | 1 - 2 | 417 | 80 | ||
BC007815.2 | - | 1 - 2 | 541 | 4299794 | 80 | |
BC008634.1 | - | 1 - 2 | 565 | 3463813 | 80 | |
BC020272.1 | - | 1 - 2 | 584 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002395.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01036979.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036979.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036971.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036971.hg.1 | ENST00000374131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036971.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036971.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036974.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036974.hg.1 | ENST00000374131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036974.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036974.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036978.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036978.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036978.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036972.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036972.hg.1 | ENST00000374131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036972.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036972.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036973.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036973.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036973.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01036975.hg.1 | TC01002395.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01036975.hg.1 | ENST00000374135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01036975.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199590.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199597.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199597.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199597.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199597.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199597.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374133.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000461282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | ENST00000477418.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013408.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199592.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199592.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199592.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199592.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199592.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199593.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199594.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199598.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199598.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199598.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | ENST00000374135.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | ENST00000431781.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | NM_018066.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100199591.hg.1 | TC0100013408.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 148072 | - | - |
