Assay Details
| Gene Symbol: CHST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX282540.1 | - | 3 - 4 | 278 | 98 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000336257.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | ENST00000565039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011567.hg.1 | NM_024533.4 | Refseq |

CHST5
Genomic Map